Learn More
Vileda Professional™ CE Sweeper and Pan™
The CE Sweeper and Pan™ is ideal for ergonomic and efficient liquid removal. The system is hygienic, lightweight, and flexible. Perfect for dry or damp conditions.
175.00 € - 316.00 €
Specifications
For Use With (Equipment) Janitorial
View More Specs
Products 2
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
17872476
View Documents
Vileda Professional™
119908
Dustpan Squeegee Sweeper
175.00 €
Pack of 5
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
17862476
View Documents
Vileda Professional™
119914
Open Lobby Dustpan
316.00 €
Pack of 5
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
Description
Description
  • Sweep up broken glass in your cleanroom quickly and easily
  • No need to change gloves when an item is dropped on the floor
  • Keeps workers from touching the floor within the controlled environment
  • Can be sprayed or wiped down with common disinfectants
  • Stands up easily for storage or can be hung
  • Adjustable bag holder for different size collection bags
Specifications
Specifications
Janitorial
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Vileda Professional™ CE Sweeper and Pan™