missing translation for 'onlineSavingsMsg'
Learn More

Vileda Professional™ CE UltraSpeed Pro™ Frame

The most effective and easy to use bucket and wringer system for use within Controlled Environments!

Brand:  Vileda Professional™ 157777

333.45 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.


Product Code. 17802506

  • 378.00 € / Pack of 10
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Demands on effectiveness, cost, ease of handling and staff training is constantly increasing. Whatever the cleaning requirements, the CE UltraSpeed Pro™ Double Bucket System will dramatically increase speed and cleaning performance while decreasing total costs and providing versatility and ergonomics.

  • Lightweight, durable and autoclavable frame
  • 40cm/16in breakaway frame can be used with all UltraSpeed Pads
  • Frame includes a modern swivel joint with integrated ball catch making it easy to clean vertical surfaces
  • Easy and fast rinsing of mops without removal of frame
  • Stainless steel springs
Specifications

Specifications

Blue
Autoclavable
119966, 157775, 157779
16 in.
10/Cs.
ISO 4 to 9/GMP A/B, C, D on Floors, Walls and Ceilings
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.