missing translation for 'onlineSavingsMsg'
Learn More
Cefotetan, 95+%, Thermo Scientific Chemicals Product Code.: 15519483

Cefotetan, 95+%, Thermo Scientific Chemicals

Product Code. 15519483
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15519483

Brand: Thermo Scientific Acros 459492500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69712-56-7
Molecular Formula C17H17N7O8S4
Molecular Weight (g/mol) 575.63
InChI Key SRZNHPXWXCNNDU-RHBCBLIFSA-N
Synonym cefotetan, cefotetanum, apacef, cefotetanum inn-latin, 6r,7s-7-4-2-amino-1-carboxy-2-oxoethylidene-1,3-dithietan-2-yl carbonyl amino-7-methoxy-3-1-methyl-1h-tetrazol-5-yl sulfanyl methyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid, cefotan tn, 6r,7s-7-4-carbamoylcarboxymethylene-1,3-dithiethane-2-carboxamido-7-methoxy-3-1-methyl-1h-tetrazol-5-yl thio methyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid, 7beta-4-2-amino-1-carboxy-2-oxoethylidene-1,3-dithietan-2-yl carbonyl amino-7alpha-methoxy-3-1-methyl-1h-tetrazol-5-yl sulfanyl methyl-3,4-didehydrocepham-4-carboxylic acid, 6r,7s-7-4-1-amino-3-hydroxy-1,3-dioxopropan-2-ylidene-1,3-dithietane-2-carbonyl amino-7-methoxy-3-1-methyltetrazol-5-yl sulfanylmethyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acidShow MoreShow Less
PubChem CID 53025
IUPAC Name (6R,7S)-7-[[4-(2-amino-1-carboxy-2-oxoethylidene)-1,3-dithietane-2-carbonyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acidShow MoreShow Less
SMILES CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)C4SC(=C(C(=O)N)C(=O)O)S4)OC)SC2)C(=O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: 0.51 mg/mL. Other solubilities: slightly soluble in methanol
Formula Weight 575.63
Percent Purity 95+%
Chemical Name or Material Cefotetan
compliance-icons
Product Identifier
  • Cefotetan
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cefotetan, 95+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.