Learn More
Hughes Ceiling Mounted Indoor Unheated Safety Shower
Ceiling mounted unheated emergency safety shower. Ideal for use where space is at a premium, this general purpose shower is suitable for use in all applications not subject to the possibility of freezing or overheating.
Brand:  Hughes HSHV0023GS
Product Code. 17772358
  • 537.00 € / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ceiling mounted unheated emergency safety shower
  • Ideal for use where space is at a premium, this general purpose shower is suitable for use in all applications not subject to the possibility of freezing or overheating
  • Fast easy grip pull handle for ease of use in an emergency
Specifications
Specifications
Stainless Steel
Ceiling
Safety Shower