Learn More
R&DSystems™ Cell Freezing Medium - II
Cell Freezing Medium-II contains 90% Fetal Bovine Serum and 10% DMSO (dimethyl sulfoxide). This cryopreservation medium is supplied as a ready-to-use solution.
Brand:  R&DSystems™ B71295
Product Code. 18412050
  • 78.00 € / 50mL
Explore more special offers
Specifications
-5 to -20 °C, protected from light
Cell Culture
0.2 μm filtered
50 mL