Learn More
Argos Technologies™ Cell-Line Adhere Bioreactor Flask
Argos Technologies™ Cell-Line Adhere Bioreactor Flasks are ideal for monoclonal antibodies, and expressed recombinant cellular protein production
Brand:  Argos Technologies™ 24406-55
Product Code. 16334358
  • 1205.00 € / Pack of 3
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • These bioreactor flasks are viable in vitro alternatives to using mouse ascites, standard cell culture, or tissue culture products
  • Bioreactors utilize innovative membrane technology to separate the cell cultivation area from the media chamber
  • Cells and cellular-secreted products with a MWCO over 10kD are retained between two non-cytotoxic membranes
  • The upper semi-permeable membrane allows continuous nutrient diffusion and waste elimination
  • The lower membrane provides direct oxygenation and ensures optimum gas exchange
  • Separate ports give access to either media or cell cultivation chamber allowing easy change of expended medium without interference of cell growth, function, or gas exchange
  • Developed for the expression of biomolecules in anchorage dependent cell lines
  • The observed product concentrations in the cell compartment are in the range of 1-5 mg/ml and are often several hundred times higher than what is obtained in either roller bottles or tissue culture flasks with the same cell type
  • Ideal for use with CHO, BHK, HEP2 or any other adherent cell line, CELLine Adhere is available in a one liter format
  • The flask is reusable and greatly reduces the downstream processing cost of concentration and purification
  • Serum usage is also reduced as a result of the compartmentalized chambers in the flask resulting is a low cost bioreactor for the production of expressed biomolecules
Specifications
Specifications
1000 mL
Polystyrene
0.2 μm Bottom Vent Membrane Barrier
Sterile