missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Cell Proliferation Kit, BioTrak™ ELISA Kit and Components

Provides a precise, fast and simple colorimetric assay to quantitate proliferating cells cultured in a 96 well microplate

Brand:  Cytiva RPN202

 View more versions of this product

Product Code. 10004694

  • 1074.00 € / 4mL
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket

Includes: The cell proliferation kit includes:
Labeling reagent, Nuclease, Anti-5-bromo-2'-deoxyuridine monoclonal antibody, Peroxidase anti-mouse IgG2a, Peroxidase anti-mouse IgG2a diluent, Substrate/intensifier

Cel proliferation Biotrak ELISA kit includes:
BrdU labelling reagent; Fixative; Blocking reagent; Peroxidase-labelled anti-BrdU; Antibody dilution solution; Wash buffer; Substrate solution

Cell proliferation labelling reagent and Anti-BrdU antibody are available separately
This item is not returnable. View return policy

Description

Description

  • Recommended cell number: 10e6-10e7 cells/mL (Cell proliferation kit) 10e2 cells/well-10e5 cells/well (Biotrak ELISA kit)
  • Required time: 2-3 hour post-labeling (labeling 1-2 hours)
  • Antibody and labeling reagent available separately for ELISA and ICC systems
  • Suitable for in vivo and in vitro tissue labeling in addition to cultured cells (ELISA format)
  • Non radioactive protocols provide sensitivity comparable to autoradiographic methods
  • Cell proliferation kit avoids the need to chemically denature DNA. Cellular morphology is preserved allowing multiple labeling experiments
  • 5-fluoro-2'-deoxyuridine is used to inhibit thymidilate synthetase increasing BrdU incorporation

When ordering the Cell Proliferation Kit (RPN20), you must also order DAB REAGENT OF RPN20/RPN1174 (RPNK8888)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

BrdU
130 μL
Immunocytochemistry
Anti-5-2'-deoxyuridine Monoclonal Antibody Nuclease
2°C to 8°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1

SDS
Documents

Documents

Certificates
Special Offers

Special Offers