Learn More
BD Biosciences™ Cell Viability Kit with Liquid Counting Beads (RUO/GMP)
Suitable for mammalian or microbial cell-based flow cyometry analyses.
Marca: BD Biosciences™ 349480
Detalles adicionales : Weight : 0.05710kg
Descripción
- Rapid counting of live/dead bacteria or other microbial cells.
- Efficacy of bacterial disinfectant.
- Viability of yeast during fermentation.
- Viability and concentration of cells in culture.
- Viability and concentration of cells before staining for flow cytometric analysis.
- Mammalian or microbial cells.
- Viability and count of cells in bioreactors.
- Viability of sperm for research studiesiability in cell preparations containing debris.
- Contents: Thiazole Orange solution, propidium iodide solution, liquid counting beads.
Especificaciones
The kit contains 1 vial of 500 μL 42 μmol/L TO in dimethyl sulfoxide (DMSO) and 1 vial of 500 μL 4.3 mmol/L PI in water. Liquid Counting Beads are supplied as 1 vial of 10 mL of fluorescent microspheres in buffer with 0.1% sodium azide. | |
Thizole orange, propidium iodide, liquid counting beads. | |
Stable for the period shon on the bottle label when stored as directed. | |
100 Tests |