Learn More
BD Biosciences™ Cell Viability Kit (RUO/GMP)
Suitable for mammalian or microbial cell-based flow cyometry analyses.
Brand:  BD Biosciences™ 349483
Additional Details : Weight : 0.00500kg
Product Code. 15746402
  • 366.00 € / 1 set
Estimated Shipment: 22-07-2024
to see stock.
Description
Description
  • Rapid counting of live/dead bacteria or other microbial cells.
  • Efficacy of bacterial disinfectant.
  • Viability of yeast during fermentation.
  • Viability and concentration of cells in culture.
  • Viability and concentration of cells before staining for flow cytometric analysis.
  • Mammalian or microbial cells.
  • Viability and count of cells in bioreactors.
  • Viability of sperm for research studiesiability in cell preparations containing debris.
  • Contents: Thiazole Orange solution, propidium iodide solution, liquid counting beads.
Specifications
Specifications
The kit contains 1 vial of 500 μL 42 μmol/L TO in dimethyl sulfoxide (DMSO) and 1 vial of 500 μL 4.3 mmol/L PI in water.
Thizole orange, propidium iodide
Stable for the period shon on the bottle label when stored as directed.
100 Tests