Learn More
Corning™ CellCube™ Culture System Clean Room Cart with Tray
The CellCube™ Culture System Clean Room Cart is a device designed for simplified installation, filling, seeding, operation and harvesting of up to four 100-layer CellCube™ modules.
Brand:  Corning™ 92341
Product Code. 17299109
  • 24975.00 € / Each
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stainless steel
  • Autoclavable
  • Mobile rack that can hold upto 4 CellCube™ modules in a small footprint
  • Modules can be rotated 360 degrees
  • Adjustable wheelbase can be sized to fit a wide selection of tracks found in incubation or environmental chambers making it a versatile product
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stainless Steel
CellCube™ modules