missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CellCube™ Systems with Corner Clips
BIOPHARMAGREENER_CHOICE

The CellCube™ module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment. It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates. Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth.

2575.00 € - 3465.00 €

Specifications

Capacity (Metric) 6 L
Includes Corner clips
Shape Square
Graduated No
Sterility Sterile
View More Specs

Products 2
Product Code Brand Surface Treatment Price Quantity & Availability  
Product Code Brand Surface Treatment Price Quantity & Availability  
17310230
BIOPHARMA
View Documents
Corning™
3032
CellBIND
3465.00 €
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
17279109
BIOPHARMAGREENER_CHOICE
View Documents
Corning™
3264
Tissue Culture-treated
2575.00 €
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
Description

Description

CellCube™ modules provide:

  • a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates.
  • 85,000 cm2 polystyrene growth surface optimally treated with the Corning™ CellBIND™ surface for the growth of attachment dependent cells.
  • a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen.
  • an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment.

Corning™ CellBIND™ Surface Benefits:

  • Improves cell attachment leading to increased cell growth and yields
  • Results in more consistent and even cell attachment
  • Helps adapt cells to reduced serum or serum-free conditions
  • Reduces premature cell detachment from confluent cultures
  • May eliminate the need for tedious, time-consuming, expensive and low stability biological coatings
  • Not a coating, requires no refrigeration or special handling and is stable at room temperature
Specifications

Specifications

6 L
Square
Sterile
100
Corner clips
No
85,000 cm2
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.