missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellEvent™ Caspase 3/7 Green Ready Flow™ Reagent

Product Code. 15945920
Click to view available options
Quantity:
120 assays
Unit Size:
120 tests
This item is not returnable. View return policy

Product Code. 15945920

Brand: Invitrogen™ R37167

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CellEvent™ Caspase-3/7 Green Ready Flow™ Reagent enables the flow cytometric detection of activated caspase-3 and caspase-7 in apoptotic cells.

The product includes 2 reagents: the novel fluorogenic substrate CellEvent™ Caspase-3/7 Green Detection Reagent and SYTOX™ AADvanced™ Dead Cell Stain.

• Stable at room temperature
• Convenient, ready-to-use format—no need to dilute, weigh, or pipette
• Exceptionally bright green fluorescence upon binding to nucleic acids
• Rapid staining of apoptotic cells without wash steps

CellEvent Caspase-3/7 Green Ready Flow Reagent is a cell-permeant reagent that detects caspase 3/7 activity using flow cytometry. Add 2 drops from the dropper bottle directly into 1 x 106 cells, incubate, and analyze.

CellEvent Caspase-3/7 Green Ready Flow Reagent consists of a four-amino acid peptide (DEVD) conjugated to a nucleic acid-binding dye. During apoptosis, caspase-3 and caspase-7 proteins are activated and able to cleave the caspase 3/7 recognition sequence encoded in the DEVD peptide. Cleavage of the recognition sequence and binding of DNA by the reagent labels the apoptotic cells with a bright, fluorescent signal having absorption/emission maxima of ∼502/530 nm. When used together with the SYTOX AADvanced Dead Cell Stain, apoptotic cells can be easily discriminated from live and necrotic cells.

Because no single parameter defines apoptosis in all systems, we strongly suggest using a combination of different measurements for reliable detection of apoptosis.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 120 assays
Format Dropper Bottle
Product Type Caspase 3/7 Reagent
Content And Storage 4 dropper bottles, store at room temperature
Excitation/Emission 504/523
Color Green
Shipping Condition Room Temperature
Sub Cellular Localization Nucleus
Product Line CellEvent™ Ready Flow
Label or Dye CellEvent™ Caspase 3/7 Green, SYTOX AADvanced Dead Cell Stain
Label Type Other Label(s) or Dye(s)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.