missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ DMEM (Dulbecco's Modified Eagle's Medium)
Corning™ DMEM (Dulbecco's Modified Eagle's Medium) support cell culture growth and substitution of Pyridoxine HCl for Pyridoxal ensures improved stability over original formulation. Products undergo a variety of tests to determine functionality and toxicity using growth-promotion analysis.
Brand: Corning™ 17-205-CV
125.90 EUR valid until 2024-12-31
Use promo code "21883" to get your promotional price.
This item is not returnable.
View return policy
Description
- Substitution of Pyridoxine HCl for Pyridoxal ensures improved stability over the original formulation
- DMEM is a modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
- Original formulation was used to culture embryonic mouse cells and has since been modified to support variety of normal and transformed cells, such as primary cultures of mouse and chicken cells
- Formulations can differ in amount or absence of components such as glucose, sodium pyruvate, sodium bicarbonate and L-glutamine
- Tested to determine pH and osmolality
- Low levels of endotoxin, and absence of contamination
- Tested for bacterial endotoxin using the Limulus Amoebocyte Lysate (LAL) Chromogenic Assay
- Osmolality: 335±mOsm/kg H2O
- Mycoplasma: Tested Negative
- Sterility: Pass
Specifications
2 to 8°C | |
Liquid | |
Glucose, Sodium Pyruvate | |
<0.25EU/mL | |
To assess growth promotion and cytotoxicity characteristics |
1 X | |
DMEM Media | |
No Glutamine, No Phenol Red | |
Inorganic Salts: 200mg/L CaCl2 (anhydrous), 0.1mg/L Fe(NO3)3.9H2O, 400mg/L KCl, 97.7mg/L MgSO4 (anhydrous), 6400mg/L NaCl, 125mg/L NaH2PO4.H2O, 3700mg/L NaHCO3; Amino Acids: 84mg/L L-Arginine. HCl, 62.57mg/L L-Cystine.2HCl, 30mg/L Glycine, 42mg/L L-Histidine.HCl.H2O, 104.8mg/L L-Isoleucine, 104.8mg/L L-Leucine, 146.2mg/L L-Lysine. HCl, 30mg/L L-Methionine, 66mg/L L-Phenylalanine, 42mg/L L-Serine, 95.2mg/L L-Threonine, 16mg/L L-Tryptophan, 103.79mg/L L-Tyrosine.2Na.2H2O, 20mg/L L-Valine; Vitamins: 4mg/L D-Calcium pantothenate, 4mg/L Choline chloride, 4mg/L Folic Acid, 7.2mg/L i-Inositol, 4mg/L Nicotinamide, 4mg/L Pyridoxine. HCl, 0.4mg/L Riboflavin, 4mg/L Thiamine. HCl; Other: 4500mg/L D-Glucose, 110mg/L Sodium pyruvate; Added: 584mg/L L-glutamine Powder, 20 mL/L 200 mM Solution. | |
6 x 500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction