missing translation for 'onlineSavingsMsg'
Learn More

Corning™ cellgro™ MEM
BIOPHARMA

Brand:  Corning™ 15-010-CM

 View more versions of this product

Product Code. 15383611

  • 249.99 € / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

To assess growth promotion and cytotoxicity characteristics, Corning Classical Media products undergo a variety of tests to determine functionality and toxicity using growth-promotion analysis. Product performance is measured by harvest-to-plant ratios or fold increase according to predetermined specifications. Liquid and powder classical media products are also tested to determine pH and osmolality. Classical media powders are also tested for residual moisture. The pH and osmolality tests are performed according to specific procedures in which all equipment is calibrated using standards traceable to the National Institute of Standards and Technology. Biological testing of Corning liquid classical media products guarantees low levels of endotoxin, mycoplasma, and an absence of contamination. Endotoxins: classical media products are tested for bacterial endotoxin using the Limulus Amoebocyte Lysate (LAL) Chromogenic Assay. For release, each lot of standard liquid classical media must have an endotoxin level of less than 0.25 EU/mL. Classical media powder products must have an endotoxin level of less than 0.25 EU/mL when reconstituted to formula weight with low endotoxin water. All Corning liquid classical media products are tested for sterility and liquid classical media products are tested for mycoplasma off-site using a large-volume method.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 X
To assess growth promotion and cytotoxicity characteristics
<0.25EU/mL
2 to 8°C
Minimum Essential Medium (MEM)
1 L
Inorganic Salts: 200.00mg/L CaCl2 (anhydrous), 400.00mg/L KCl, 97.70mg/L MgSO4 (anhydrous), 6,800.00mg/L NaCl, 140.00mg/L NaH2PO4.H2O, 2,200.00mg/L NaHCO3; Amino Acids: 126.40mg/L L-Arginine. HCl, 31.20mg/L L-Cystine.2HCl, 41.90mg/L L-Histidine.HCl.H2O, 52.50mg/L L-Isoleucine, 52.50mg/L L-Leucine, 72.50mg/L L-Lysine. HCl, 15.00mg/L L-Methionine, 32.50mg/L L-Phenylalanine, 47.60mg/L L-Threonine, 10.00mg/L L-Tryptophan, 51.90mg/L L-Tyrosine.2Na.2H2O, 46.80mg/L L-Valine; Vitamins: 1.00mg/L D-Calcium pantothenate, 1.00mg/L Choline chloride, 1.00mg/L Folic acid, 2.00mg/L i-Inositol, 1.00mg/L Nicotinamide, 1.00mg/L Pyridoxine. HCl, 0.10mg/L Riboflavin, 1.00mg/L Thiamine. HCl; Other: 1,000.00mg/L D-Glucose, 10.00mg/L Phenol red. Na.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ cellgro™ MEM > Capacity: 1L; Product Type: With Earle’s Salts

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.