missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ cellgro™ Tryptic Soy Broth

Brand: Corning™ 46-060-CM
This item is not returnable.
View return policy
Description
Tryptic soy broth is offered for use in sterility testing and validation in the manufacture of a liquid product. The use of this medium in place of the product intended for production allows for the assessment of the capability of an aseptic manufacturing process. It is a sterile-filtered, ready-to-use solution for the simulation of aseptic filling of liquids and sterility testing. Due to the rich nutrient base, TSB may be used in the cultivation of fastidious microorganisms. This growth medium is free of animal-sourced components, making it ideal for testing the sterility of animal-free biopharmaceutical processes.Order Info
dimQuantity

Specifications
Bacterial Growth Medium | |
Sterile |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
33824013 | Certificate of Quality | 15/03/2025 | 46-060-CM |
26924007 | Certificate of Quality | 01/01/2025 | 46-060-CM |
17724026 | Certificate of Quality | 23/10/2024 | 46-060-CM |
35022007 | Certificate of Analysis | 02/06/2023 | 46-060-CM |
26422007 | Certificate of Analysis | 31/03/2023 | 46-060-CM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ cellgro™ Tryptic Soy Broth > Capacity: 1L
Spot an opportunity for improvement?Share a Content Correction