Learn More
Thermo Scientific™ CellInsight™ CX7 LZR High Content Analysis Platform and Store Standard Edition (SE)
CellInsight CX7 LZR High Content Analysis Platform and Store Standard Edition (SE)
Brand: Thermo Scientific™ CX7B1112LZR
This item is not returnable.
View return policy
Description
The Thermo Scientific CellInsight CX7 LZR High Content Analysis (HCA) Platform is a fast, laser-based, automated cellular imaging and analysis platform designed for quantitative microscopy. This particular configuration includes a CellInsight CX7 LZR Reader, HCS Studio Cell Analysis Software, and Store Image and Database Management Software to visualize and share high content data and images. Two days of onsite training and a one-year warranty and application support are also included.The CellInsight CX7 LZR HCA Platform offers a more powerful and integrated way to develop and automate high content assays using a combination of 5-color bright-field and 7-color fluorescence imaging in confocal or wide-field modes. With laser illumination, the Cell Insight LZR HCA Platform enables researchers to image faster in both wide-field and confocal mode.
Features of the CellInsight CX7 HCA Platform with Store Image and Database Management Software:
• 7-channel fluorescent imaging with laser illumination
• 5-channel bright-field imaging
• 7-channel confocal imaging
• Software and laser-based autofocus for consistent scan times
• Configured to accept optional modules for fully automated plate handling and live cell on-board environmental control
Capabilities
• Addresses hundreds of assay types with flexible software and hardware tools
• Scan and analyze microscope slides or most SBS-standard format microplates
• Use integrated HCS Studio software for instrument control and data analysis
• Achieve the fastest time to results by using intelligent acQuisition (iQ) real-time analysis feedback
• Compatible with Molecular Probes fluorescent detection reagents
Hardware and software
• High-output laser excitation light
• Broad reagent options with seven-color excitation from violet to near IR for assay multiplexing
• Dual mode focusing using Software and Laser Autofocus methods to address both labelled and unlabeled sample types
• Multi-mode imaging allows for wide-field, bright-field, and confocal imaging in one run
• HCS Studio Cell Analysis Software is an integrated instrument control, visualization, and analysis software package for novice to advanced users
• Scalable with Store Image and Database Software (included) and optional Orbitor RS Microplate handler (not included)
Specifications
CellInsight | |
Fluorescence | |
Room Temperature |
Assorted Colors | |
Less hazardous, less waste and use of fewer resources |