missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Cellmaxx™ Fetal Bovine Serum, New Zealand Origin
Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
- This CellMaxx FBS comes from the pristine grasslands of New Zealand where the cattle are tracked throughout their life and has never had BSE or FMD, meaning it has the lowest possible viral risk.
- It is produced to the highest standards to ensure product performance and viral safety while maintaining minimal lot-to-lot variability.
- Each lot is tested in MBDK, hybridoma, MRC-5 and Vero cell lines for growth promotion.
- The absence of ten viruses is confirmed according to 9CFR.
Specifications
Specifications
| Form | Liquid |
| Quantity | 500 mL |
| Endotoxin Concentration | 5 IU/mL |
| Hemoglobin Concentration | 18 mg/mL |
| IgG levels | 400 mg/L |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction