Learn More
Cellpath System II HEX™ Cassette Lid
For use in place of plastic lid on System II HEX tissue processing cassettes. CellPath Tissue Processing System II HEX Cassette Lid is made of stainless-steel.
Brand:  Cellpath EAW-0114-08A
Product Code. 30226268
  • 268.70 € / Pack of 25
Explore more special offers
Specifications
25/Pk.
Stainless steel
System II HEX Cassette lid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cellpath System II HEX™ Cassette Lid > System II HEX Cassette lid