missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CellTrace™ Calcein Red-Orange, AM - Special Packaging
CellTrace™ Calcein Red-Orange, AM - Special Packaging
Brand: Invitrogen™ C34851
568.10 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
CellTrace calcein red-orange AM is a cell-permeant dye that can be used to determine cell viability in most eukaryotic cells. Unlike calcein AM (C-1430, C-3099, C-3100), CellTrace calcein red-orange AM is intrinsically fluorescent; thus, an additional wash step may be necessary to minimize background fluorescence from dye that is not taken up by cells. However, CellTrace calcein red-orange (excitation/emission maxima 577/590 nm) is well-retained by live cells that possess intact plasma membranes, and consequently it is a useful cell tracer and indicator of cell viability.Specifications
Cell-permeant | |
Calcein, Red-Orange, AM - Special Packaging | |
Fluorescence Microscope | |
Other Label(s) or Dye(s) | |
577 nm | |
CellTrace™ | |
Cell Tracker Compounds, Cell Labeling Reagents | |
Esterase |
Store in freezer at -5°C to -30°C and protect from light. | |
Cell Tracing, Cell Tracker | |
Dye | |
590 nm | |
Lyophilized | |
20 x 50 μg | |
Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction