missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ CellTrace™ Calcein Violet, AM, for 405 nm excitation - Special Packaging
CellTrace™ Calcein Violet, AM, for 405 nm excitation - Special Packaging
Brand: Molecular Probes™ C34858
This item is not returnable.
View return policy
Description
CellTrace™ calcein violet AM is an optimal dye for distinguishing live cells through the presence of ubiquitous intracellular esterase activity. The enzymatic conversion of the virtually non-fluorescent cell-permeant calcein violet AM to the intensely fluorescent calcein violet (ex/em 400/452 nm) can be easily detected with the violet laser, allowing other laser lines to be used for conventional fluorochromes.View a selection guide for all Nonfixable Viability Dyes for Flow Cytometry.
Specifications
Contains 20 vials of calcein violet AM (25 μg per vial). Store in freezer (-5 to -30°C) and protect from light. | |
calcein violet | |
Flow Cytometer | |
Fluorescence | |
Cell Tracker Compounds, Cell Labeling Reagents | |
Dye |
400⁄452 | |
Tube(s) | |
1000 Reactions | |
CellTrace™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.