Learn More
Biodesign™ Cellulose Dialysis Tubing Roll
Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies.
127.00 € - 377.00 €
Specifications
Length (Metric) 30.5 m
Color Transparent
View More Specs
Products 13
Product Code Brand Capacity (Metric) MWCO Diameter (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) MWCO Diameter (Metric) Price Quantity & Availability  
12787466
View Documents
Biodesign™
D002
1.91 mL/cm 14 kDa 15.5 mm
187.00 €
Each
12777466
View Documents
Biodesign™
D001
0.32 mL/cm 14 kDa 6.4 mm
271.00 €
Each
12717476
View Documents
Biodesign™
D008
19.13 mL/cm 14 kDa 49.5 mm
333.00 €
Each
12707476
View Documents
Biodesign™
D006
6.45 mL/cm 14 kDa 28.7 mm
190.00 €
Each
12777476
View Documents
Biodesign™
D100
0.32 mL/cm 8 kDa 6.4 mm
210.00 €
Each
12787476
View Documents
Biodesign™
D102
1.91 mL/cm 8 kDa 15.5 mm
184.00 €
Each
12797476
View Documents
Biodesign™
D104
3.57 mL/cm 8 kDa 21.3 mm
175.00 €
Each
12707486
View Documents
Biodesign™
D106
6.45 mL/cm 8 kDa 28.7 mm
202.00 €
Each
12717486
View Documents
Biodesign™
D108
19.13 mL/cm 8 kDa 49.5 mm
299.00 €
Each
12777486
View Documents
Biodesign™
D302
1.91 mL/cm 3.5 kDa 15.5 mm
350.00 €
Each
12787486
View Documents
Biodesign™
D304
3.57 mL/cm 3.5 kDa 21.3 mm
302.00 €
Each
12797486
View Documents
Biodesign™
D306
6.45 mL/cm 3.5 kDa 15.5 mm
377.00 €
Each
12797466
View Documents
Biodesign™
D004
3.57 mL/cm 14 kDa 21.3 mm
127.00 €
Each
Description
Description
Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies.
  • Seamless, thin, transparent dialysis tubing manufactured from the highest purity cellulose
  • Just wet in distilled water and use
  • Available in 30.5m rolls
  • Specially purified for life science research
  • Supplied in an airtight, moisture-proof container for maximum shelf life
  • Available in 14000Da, 8000Da, and 3500Da molecular weight cut offs
Specifications
Specifications
30.5 m
Transparent