Learn More
Macherey-Nagel™ Cellulose Extraction Thimbles
Minimizes the hands-on time involved in extraction. Macherey Nagel™ Cellulose Extraction Thimbles are used for holding solid materials from which substances are to be extracted with a suitable solvent.
Brand:  Macherey-Nagel™ 645025
 View more versions of this product
Product Code. 10422032
  • 145.00 € / Pack of 25
Explore more special offers
Description
Description
  • Manufactured from pure cellulose
  • Used in the fields of air and waste gas analysis for collecting solid particles
  • The choice of analytical chemists using Soxhlet or automated extractors
Specifications
Specifications
25 Pk.
Cellulose
180 mm
31 mm
Analysis of soil contaminants, Analysis of soil contaminants
Extraction Thimbles