missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Cellulose Membranes
Suitable for Clarification and sterilizing filtration of biological fluids . Fisherbrand™ Cellulose Membranes filters are composed of pure cellulose acetate modified to offer researchers the lowest binding filters available.
Brand: Fisherbrand™ 0454A00022
41.88 EUR valid until 2024-12-31
Use promo code "21757" to get your promotional price.
Description
- Filtration sterile of aqueous solutions (nutrient media, sera, buffers)
- Sterilizing filtration of proteins and enzymes
- Provide high throughputs
- Reduces filter changes when filtering proteinaceous solutions
- Has Receptor binding studies
- Low binding characteristics
- Made of hydrophilic membranes
- Uniform pore size structure allows consistent flow rates
- Present strength and dimension stability (130psi) and resist to 135°C
Specifications
3.1bar | |
Microfilter | |
0.45 μm | |
40mL/min./cm2 | |
25L/min./cm2 | |
100 Pack | |
Yes |
White | |
Cellulose Acetate | |
Non-sterile | |
Sterilizing filtration of aqueous solutions (Nutrient media, Buffers and sera), sterilizing filtration of Proteins and enzyme, receptor binding studies | |
Hydrophilic | |
Plain Disc | |
25 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction