missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Cellulose Membranes
Suitable for Clarification and sterilizing filtration of biological fluids . Fisherbrand™ Cellulose Membranes filters are composed of pure cellulose acetate modified to offer researchers the lowest binding filters available.
Brand: Fisherbrand™ 0452A00022
Description
- Filtration sterile of aqueous solutions (nutrient media, sera, buffers)
- Sterilizing filtration of proteins and enzymes
- Provide high throughputs
- Reduces filter changes when filtering proteinaceous solutions
- Has Receptor binding studies
- Low binding characteristics
- Made of hydrophilic membranes
- Uniform pore size structure allows consistent flow rates
- Present strength and dimension stability (130psi) and resist to 135°C
Specifications
4bar | |
Micro filtration | |
0.2 μm | |
18.5mL/min./cm2 | |
Hydrophilic | |
Plain Disc | |
25 mm |
White | |
Cellulose Acetate | |
Non-sterile | |
Sterilizing filtration of aqueous solutions (Nutrient media, Buffers and sera), sterilizing filtration of Proteins and enzyme, receptor binding studies | |
100 Pack | |
Yes |