missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Cellulose Membranes
Suitable for Clarification and sterilizing filtration of biological fluids . Fisherbrand™ Cellulose Membranes filters are composed of pure cellulose acetate modified to offer researchers the lowest binding filters available.
45.40 € - 99.75 €
Specifications
Color | White |
---|---|
Filter Type | Microfilter |
Material | Cellulose Acetate |
Sterility | Non-sterile |
For Use With (Application) | Sterilizing filtration of aqueous solutions (Nutrient media, Buffers and sera), sterilizing filtration of Proteins and enzyme, receptor binding studies |
Product Code | Brand | Bubble Point | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bubble Point | Diameter (Metric) | Price | Quantity & Availability | ||||
11715315
|
Fisherbrand™
0454A00022 |
3.1bar | 25 mm |
45.40 €
Pack of 100 |
Estimated Shipment: 09-01-2025
Log in to see stock. |
||||
11725315
|
Fisherbrand™
0454A00023 |
3.1bar | 47 mm |
94.80 €
Pack of 100 |
Estimated Shipment: 21-01-2025
Log in to see stock. |
||||
11705315
|
Fisherbrand™
0452A00023 |
4bar | 47 mm |
99.75 €
Pack of 100 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
11795305
|
Fisherbrand™
0452A00022 |
4bar | 25 mm |
45.40 €
Pack of 100 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
Description
- Filtration sterile of aqueous solutions (nutrient media, sera, buffers)
- Sterilizing filtration of proteins and enzymes
- Provide high throughputs
- Reduces filter changes when filtering proteinaceous solutions
- Has Receptor binding studies
- Low binding characteristics
- Made of hydrophilic membranes
- Uniform pore size structure allows consistent flow rates
- Present strength and dimension stability (130psi) and resist to 135°C
Specifications
White | |
Cellulose Acetate | |
Sterilizing filtration of aqueous solutions (Nutrient media, Buffers and sera), sterilizing filtration of Proteins and enzyme, receptor binding studies | |
100 Pack | |
Yes |
Microfilter | |
Non-sterile | |
Hydrophilic | |
Plain Disc |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title