missing translation for 'onlineSavingsMsg'
Learn More

LABPLAS Cellulose Sani-Sponge with Letheen Broth

Cellulose sponge for sampling surfaces, with buffer to neutralize disinfectant.

258.00 € - 301.00 €

Specifications

Product Type Hygiene Sponge Sampling Kit
Media Type Neutralizers
Sterility Sterile
View More Specs

Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
17906689
View Documents
LABPLAS
KSS-67110-LT
None
258.00 €
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
17926509
View Documents
LABPLAS
KSS-67115-LT
Gloves
301.00 €
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Cellulose Sani-Sponge
  • Saturated with letheen broth
  • Dimensions: 160 x 229 mm
  • Holds: 600 mL
  • Option to include gloves with kit
Specifications

Specifications

Hygiene Sponge Sampling Kit
Neutralizers
Sterile
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
LABPLAS Cellulose Sani-Sponge with Letheen Broth

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.