missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LABPLAS Cellulose Sani-Stick with Buffered Peptone Water
Cellulose sponge with stick for sampling surfaces, with buffer to neutralize disinfectant.
271.00 € - 326.00 €
Spécification
Product Type | Hygiene Sponge Sampling Kit |
---|---|
Media Type | Neutralizers |
Sterility | Sterile |
Code produit | Marque | Includes | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Includes | Prix | Quantité et disponibilité | |||||
17906649
|
LABPLAS
KSS-67310-BPW |
None |
271.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17956749
|
LABPLAS
KSS-67315-BPW |
Gloves |
326.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Cellulose Sani-Stick
- Saturated with buffered peptone water
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Option to include gloves with kit
Spécification
Hygiene Sponge Sampling Kit | |
Neutralizers | |
Sterile |