missing translation for 'onlineSavingsMsg'
Learn More

Scotch-Brite™ Cellulose Scouring Pad 770

Abrasive power for use on a variety of surfaces

Brand:  Scotch-Brite™ 7100157792

Product Code. 17188770

  • 772.00 € / Pack of 240
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sponge
88 x 131 mm
Cellulose
131 mm
Blue
88 mm
240 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Scotch-Brite™ Cellulose Scouring Pad 770 > Blue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.