missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cellulose triacetate

Product Code. 10124340
Click to view available options
Quantity:
250 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-09-3
Molecular Formula (C24H32O16)n
MDL Number MFCD00132680

Specifications

Color White to Yellow
Physical Form Pellets or Rods
Merck Index 10, 1926
Solubility Information Solubility in water: .. Other solubilities: soluble in chloroform, methylene dichloride and, tetrachloroethane
Quantity 500 g
SMILES CC(=O)OC[C@H]1O[C@@H](O[C@@H]2[C@@H](COC(C)=O)OC(-*)[C@H](OC(C)=O)[C@H]2OC(C)=O)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1O-*
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material Cellulose triacetate
Product Identifier
  • Cellulose triacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.