missing translation for 'onlineSavingsMsg'
Learn More

Celpure™ P65, Meets USP/NF Testing Specifications, Honeywell Fluka™

Product Code. 15625750
Click to view available options
Quantity:
250 g
Packaging:
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

meets USP/NF testing specifications

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Density 0.10 to 0.26 g/cm3 (wet density)
Color Gray
Quantity 250 g
pH 5 to 8
CAS 61790-53-2
Loss on Drying 0.01 to 0.50% at 105°C
Loss on Ignition 0.01 to 2.00% at 1000°C
Packaging Plastic Bottle
Permeability 0.04 to 0.08 D
Silicon Dioxide 98%
Molecular Formula O2Si
MDL Number MFCD00132803
UN Number NONH for all modes of transport
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES O=[Si]=O
IUPAC Name dioxosilane
Molecular Weight (g/mol) 60.083
PubChem CID 24261
ChEBI CHEBI:30563
Chemical Name or Material Celpure™ P65
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.