missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CELT-133

Selective hα1A adrenergic receptor fluorescent antagonist

Brand:  Tocris Bioscience™ 7952/10U

380.70 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18085962

  • 465.00 € / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

CELT-133 is a selective hα1A adrenergic receptor fluorescent antagonist. It shows full selectivity for α1A over α2A (Ki= 5 nM and 15% of displacement at 1 μM respectively in radioligand binding assay). CELT-133 binds equally to all subtypes of α1 adrenergic receptor. Excitation and emission maxima (λ) are 560 and 571 nm, respectively. Wavelengths are compatible with use as an acceptor dye in TR-FRET assays, together with the CoraFluor™ 1 (Cat. No. 7920) TR-FRET donor. Add 76 μL of assay buffer containing 1% of DMSO to obtain a 100 μM stock solution, and use 250 nM as a working concentration.
Specifications

Specifications

10 μg
Fluorescent Receptor Probes
95%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers