missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Centrifichem™, Technicon™ Beckman™ Polystyrene Sample Cups

Product Code. p-6858001
Click to view available options
Capacity (Metric):
0.5 mL
2.0 mL
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 11393804

Brand: Kartell™ 02511004

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Manufactured with crystal clear polystyrene for optimum visibility. Kartell™ Technicon™ Beckman™ Konelab™ Sample Cups are used for specimen analysis.

  • A range of crystal clear polystyrene auto analyser cups with conical bases
  • Stand-alone construction
  • CE-IVD approved

Specifications

Quantity 1000/pk.
Capacity (Metric) 0.5 mL
Color Clear
Product Type Sample Cup
Material Polystyrene
Disposable Single-use
For Use With (Equipment) Centrifichem, Technicon and Beckman analyzers
Height (Metric) 24.5 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.