Learn More
Eppendorf™ Steel Sleeves for Centrifuge Adapter
Designed for use with Eppendorf Rotors and Centrifuges
Brand:  Eppendorf™ 5820774002
Product Code. 13246619
  • 360.00 € / Pack of 24
Explore more special offers
Description
Description
  • Accessory for Eppendorf liquid handling equipment
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Steel Sleeves including Adapter for Rotor
36 x 15 mL Tubes
100 mm
Steel sleeves including adapter for rotor
24/Pk.
Steel
16 mm
Rotor F-35-48-17
Blood Collection Tube