Learn More
Eppendorf™ Centrifuge Adapters
Designed for use with Eppendorf Rotors and Centrifuges
85.80 € - 108.00 €
Product Code | Brand | Description | Capacity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Capacity | Price | Quantity & Availability | ||||
10592774
|
Eppendorf™
5427734007 |
Adapter, large bore | 1 x 7 to 15 mL Blood Collection Tube |
108.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10638004
|
Eppendorf™
5427732004 |
Adapter, small bore | 1 x 7 to 15 mL Blood Collection Tube |
108.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10789272
|
Eppendorf™
5702702005 |
Adapter | 1 x 15 mL Conical Tube |
85.80 €
Pack of 8 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Accessory for Eppendorf liquid handling equipment
Specifications
Rotor Adapter |