Learn More
OHAUS™ Centrifuge Rack
Adapter accessory for Centrifuge Multi FC5916R and Centrifuge Multi FC5916
Brand:  OHAUS™ 30553131
298.30 EUR valid until 2024-11-30
Use promo code "21681" to get your promotional price.
 View more versions of this product
Product Code. 16632774
Description
Description
  • Adapter for centrifuges:
    • Centrifuge Multi FC5916R
    • Centrifuge Multi FC5916
TRUSTED_SUSTAINABILITY
Specifications
Specifications
7 x 50 mL
Centrifuge Multi FC5916R and Centrifuge Multi FC5916
Depth 29 mm
RB x2