Learn More
OHAUS™ Centrifuge Rack
Adapter accessory for Centrifuge Multi FC5916R and Centrifuge Multi FC5916
Brand:  OHAUS™ 30553133
278.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16652774
  • 287.00 € / Pack of 2
Explore more special offers
Description
Description
  • Adapter for centrifuges:
    • Centrifuge Multi FC5916R
    • Centrifuge Multi FC5916
TRUSTED_SUSTAINABILITY
Specifications
Specifications
14 x 15 mL
Centrifuge Multi FC5916R and Centrifuge Multi FC5916
Depth 17 mm
FA x2