missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Centrifuge Trolley

Ergonomic, benchtop trolley for Legend RM and Multifuge 1 S-R, 1 L-R Centrifuges.

Brand:  Thermo Scientific 75000075

2036.06 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.


Product Code. 10209204

  • 2502.45 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Ergonomic, benchtop trolley for Legend RM and Multifuge 1 S-R, 1 L-R Centrifuges
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Benchtop Trolley
Legend RM and Multifuge 1 S-R, 1 L-R Centrifuges
Reusable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers