missing translation for 'onlineSavingsMsg'
Learn More

Hecht Karl™ Assistant™ Centrifuge Tubes ASTM

Cylindrical Assistant™ Centrifuge Tubes ASTM feature a conical bottom, with ASSISTENT-PERMANENT fused blue graduation marks. Manufactured from borosilicate glass 3.3, they comply with ASTM D 96 DIN 51793.

Brand:  Hecht Karl™ 40930010

Product Code. 10236951

  • 40.00 € / Each
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Cylindrical centrifuge tubes with conical bottom
  • ASSISTANT-PERMANENT fused blue graduations
  • Borosilicate glass 3.3
Specifications

Specifications

ASTM D 96 DIN 51793
Borosilicate Glass (3.3)
Cylindrical
Conical
100 mL
Centrifuge applications and general lab use
3.4 oz.
Centrifuge Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hecht Karl™ Assistant™ Centrifuge Tubes ASTM > Assistant™ Centrifuge Tubes ASTM; Capacity: 100 mL (3.4 oz.)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.