missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terracycle™ Centrifuge Tubes and Rigid Lab Plastics Zero Waste Box™

Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
Description
Use this Zero Waste Box to recycle:
- Any brand and type of rigid lab plastics: centrifuge tubes, plastic bottles, trays, vials and beakers.
Approximate waste units accepted:
- Small: 200 units
- Medium: 550 units
- Large: 1000 units
Do not send in:
- Tubes and rigid lab plastics that are contaminated with bodily fluids and materials considered hazardous or dangerous
- Autoclaved material
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
- Batteries, pressurized canisters, broken glass and medical waste
Important:
- We do not collect any hazardous waste containing substances with toxic, reactive, corrosive, infectious, flammable, explosive properties or harmful to human health and the environment.
- Please make sure that you do not mix hazardous waste with non-hazardous waste.
- As the waste producer, it is your responsibility to classify the waste.
Specifications
Specifications
| Capacity (Metric) | 7 kg |
| Capacity (English) | 15 lb. |
| Color | Beige, Green, Red, White |
| Dimensions (L x W x H) | 25 x 25 x 46 cm |
| Disposable | Single-use |
| Latex Free | Yes |
| Size | Small |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Material | Cardboard, Plastic |
| Show More |
Product Content Correction
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur