Learn More
Labconco™ CentriVap Aqueous System
This system includes: Benchtop Vacuum Concentrator; -50 Cold Trap; Clear Canister; Moisture Insert; Rotor; and Vacuum Tubing and 6 Clamps
Brand:  Labconco™ 7982035
Product Code. 15243463
  • 13765.00 € / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
This CentriVap Vacuum Concentration System includes the following components: Benchtop Vacuum Concentrator; -50 Cold Trap; Clear Canister 7460900; Moisture Insert 7814900; 12-13 mm Rotor 7455100; and Vacuum Tubing and 6 Clamps. 230V, 50Hz, UK plug
Specifications
Specifications
CE Marked
Benchtop Vacuum Concentrator; -50 Cold Trap; Clear Canister; Moisture Insert; 12-13 mm Rotor; Vacuum Tubing and 6 Clamps
Corrosion Resistant Steel
International
0 to 300 V
230 V 50 Hz