missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ CentriVap™ Complete Concentrator
BIOPHARMA

Comes with everything needed to start processing

22560.00 € - 27815.00 €

Specifications

Model Centrivap
View More Specs

Products 4
Product Code Brand Electrical Rating Electrical Requirements Type Price Quantity & Availability  
Product Code Brand Electrical Rating Electrical Requirements Type Price Quantity & Availability  
15263423
BIOPHARMA
View Documents
Labconco™
7315063
International/UK 230 V 50 Hz 6 A Acid Resistant with Heat Boost
27815.00 €
Each
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
 
15243423
BIOPHARMA
View Documents
Labconco™
7315062
International/UK 230 V 50 Hz 6 A Heat Boost Included
23120.00 €
Each
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
 
15233423
BIOPHARMA
View Documents
Labconco™
7315061
International/UK 230 V 50 Hz 6 A Acid Resistant without Heat Boost
27265.00 €
Each
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
 
15223423
BIOPHARMA
View Documents
Labconco™
7315060
International/UK 230 V 50 Hz 6 A Heat Boost Not Included
22560.00 €
Each
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
Description

Description

Comes with everything needed to start processing

Uses a combination of centrifugal force, vacuum and heat to speed evaporation.

Includes:

  • Vacuum Concentrator
  • -50°C Cold Trap
  • Aluminum rotor holds 40 microcentrifuge tubes and 12-13mm tubes
  • Chemical-resistant diaphragm vacuum pump
  • CentriZap Strobe Light allows user to view samples while rotor is spinning and lid is closed

Molecular biology, proteomics, genomics, cell biology, microbiology, biochemistry, drug discovery and analytical chemistry

Specifications

Specifications

Centrivap
Videos
SDS
Documents

Documents

Product Certifications