Learn More
Labconco™ CentriVap DNA Vacuum Concentrator
Speeds up the evaporation of solvents from DNA samples and other very small samples. Labconco™ CentriVap Vacuum Concentrator does not require any additional components to begin processing.
Brand:  Labconco™ 7970035
Product Code. 13426188
  • 12260.00 € / Each
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Quick-Start™ One Button Start Up saves time.
  • Quick-Stop™ System allows pump to stop within seconds. The valve opens to bleed air into the chamber, and the rotor stops.
  • Memory stores 9 user-set programs.
  • Easy-to-read LCD display prompts the user to set program parameters and displays program number, actual and set point temperatures, point heat, and run time remaining.
  • Microprocessor-controlled 300-watt heater supplies a controlled amount of heat from OFF up to a maximum of 100 ° C in one degree increments.
  • Heat and run times are set separately, each from 1 to 999 minutes so that heat may be programmed to shut down sooner than the run to protect heat sensitive samples from excessive heat exposure after the solvent has evaporated.
  • PREHEAT button
  • Microprocessor-controlled centrifugal motion is provided by a brushless motor capable of achieving up to 1725 RPM.
  • Audible alarm signals completion of set point run time.
  • Corrosion-resistant sample chamber accommodates a variety of rotors.
  • Versatile DNA Rotor is included.
  • Accommodates up to 60 each 1.5 ml and 72 each 0.5 ml microcentrifuge tubes.
  • Integrated, corrosion-resistant diaphragm pump achieves vacuum of 752mm Hg and has a pumping capacity of 34 liters per minute.
  • Automatic vacuum release valve prevents loss of sample by releasing the vacuum before the rotor stops and during power failures.
  • Large phenol-free lid seal ensures good vacuum without potential for phenol contamination.
  • Lid latch with safety sensor locks the lid in place when the rotor is turning and prevents start up when the lid is open.
  • Built-in vacuum delay prevents bumping by allowing the rotor to achieve speed before applying vacuum.
  • Two 177 milliliter glass traps protect the vacuum pump and the environment.
  • ETL listed (115 volt models)
  • CE (European Community) conformity (230 volt models)
  • Full one year warranty is provided against defects in materials and workmanship.
Specifications
Specifications
CE (European Community) conformity (230 volt models)
60 x 1.5 mL, 72 x 0.5 mL Microcentrifuge Tubes
230 V
230 V 50/60 Hz 6 A International
British (UK)
45 kg
50/60 Hz
Acryllic lid, DNA Rotor, diaphragm vacuum pump, two glass traps and all connecting tubing
Up to 100°C
1725rpm
300 w
404 x 663 x 284 mm