Learn More
Labconco™ CentriVap Mobile System
CentriVap Mobile Systems combine the separate vacuum concentrator and cold trap components into one space-saving console and include casters for portability from laboratory to laboratory. A 12-13 mm Rotor and Solvent Trap are also included. The interior can accommodate a vacuum pump.
Brand: Labconco™ 7812035
This item is not returnable.
View return policy
Description
CentriVap Mobile Systems combine the separate vacuum concentrator and cold trap components into one space-saving console and include casters for portability from laboratory to laboratory. A 12-13 mm Rotor and Solvent Trap are also included. The interior can accommodate a vacuum pump — the only additional component necessary to begin processing. 230V, 50Hz, UK plugSpecifications
UL 3101-1/61010-1, CE (European Community) conformity | |
210 lb. | |
0 to 300 V | |
230 V 50 Hz 8 A | |
38.2 in. | |
35.8 cm | |
70.9 cm | |
14.1 x 27.9 x 38.2 in. (35.8 x 70.9 x 97.0 cm) |
International | |
95.0 kg | |
Corrosion Resistant Steel | |
14.1 in. | |
97.0 cm | |
British (UK) | |
27.9 in. |
Product Content Correction
Product Title
Labconco™ CentriVap Mobile System > CentriVap Mobile System. Combines vacuum concentrator and cold trap components into space-saving console. Includes casters, rotor and solvent trap. Interior accommodates a vacuum pump.
Spot an opportunity for improvement?Share a Content Correction