Learn More
Labconco™ Proteomic CentriVap Concentrator Systems
Proteomic CentriVap Concentrator Systems include all the concentration components necessary to begin operation except for the vacuum pump.
Brand: Labconco™ 7989635
This item is not returnable.
View return policy
Description
Proteomic CentriVap Concentrator Systems include all the concentration components necessary to begin operation except for the vacuum pump. The Refrigerated CentriVap Vacuum Concentrator concentrates multiple small heat-sensitive samples, such as RNA and proteins, using centrifugal force, vacuum and controlled temperature to -4° C (25° F). The DNA Rotor accommodates up to 60 each 1.5 mL and 72 each 0.5 mL microcentrifuge tubes. The CentriVap Cold Trap reaches -50° C (-58° F) for solvent recovery and to protect the vacuum pump by trapping moisture, vapors and corrosive fumes as they evaporate from the samples. 230V, 50Hz, UK plugSpecifications
CE Marked | |
All concentration components necessary to begin operation except for vacuum pump | |
Corrosion Resistant Steel | |
British (UK) |
International | |
0 to 300 V | |
230 V 50 Hz |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Labconco™ Proteomic CentriVap Concentrator Systems > Proteomic CetriVap System, includes concentrator, rotor and cold trap
Spot an opportunity for improvement?Share a Content Correction