Learn More
Tocris Bioscience™ CEPT Cocktail Kit
Cell culture supplement for improving stem cell survival
Brand:  Tocris Bioscience™ 7991/1 KIT
293.73 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 30000439
  • 384.00 € / 1 set
Explore more special offers
Description
Description
The CEPT Cocktail Kit is an easy-to-use kit that consists of Chroman 1 (Cat. No. 7163), Emricasan (Cat. No. 7310), Polyamine Supplement (Cat. No. 7739), trans-ISRIB (Cat. No. 5284) and DMSO (Cat. No. 3176). It contains sufficient material for the preparation of 5 L of cocktail and sterile-filtered DMSO for reconstitution. The CEPT Cocktail is a cell culture supplement for improving the survival of self-renewing pluripotent stem cells and their differentiated progeny in stem cell research. It can be used for routine long-term passaging, cryopreservation, single-cell cloning, and organoid formation.
Please refer to the protocol for further information.
Specifications
Specifications
1 Kit
Compounds for Stem Cell Proliferation & Renewal