Learn More
BUCHI Ceramic Crucible
For use with B-440 Wet Digesters to aid in the ecological pre-incineration of different types of samples.
Brand:  BUCHI 026938
Product Code. 12769785
  • 116.00 € / Each
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 90mL capacity
  • White in color
  • Round in shape
Specifications
Specifications
White
Pre-incineration of samples
Crucible
Ceramic
90 mL
Round