Learn More
Thermo Scientific™ Ceramic Homogenizers for QuEChERS Kits
Use Thermo Scientific™ Ceramic Homogenizers to grind and homogenize samples for your QuEChERS sample preparation workflow.
Brand:  Thermo Scientific™ 60106-CH-2
 View more versions of this product
Product Code. 17339822
  • 49.04 € / Pack of 200
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Use Thermo Scientific™ Ceramic Homogenizers to help accelerate the homogenization process and increase recovery and reproducibility during your QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) sample preparation workflow. These homognizers can be used with all of our QuEChERS Extraction and Clean-Up kits to supplement Original 2003 Method, AOAC 2007.01 Method, EN 15662 Method, and NY/T 1380-2007 Method procedures.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ceramic Homogenizers for 2 mL Tubes, 200/Pk.
2 mL Tubes
Ceramic Homogenizer