Learn More
Thermo Scientific™ Ceramic Homogenizers for QuEChERS Kits
Use Thermo Scientific™ Ceramic Homogenizers to grind and homogenize samples for your QuEChERS sample preparation workflow.
Brand:  Thermo Scientific™ 60106-CH-50
46.65 EUR valid until 2025-03-14
Use promo code "24142" to get your promotional price.
 View more versions of this product
Product Code. 17349822
  • 49.10 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Use Thermo Scientific™ Ceramic Homogenizers to help accelerate the homogenization process and increase recovery and reproducibility during your QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) sample preparation workflow. These homognizers can be used with all of our QuEChERS Extraction and Clean-Up kits to supplement Original 2003 Method, AOAC 2007.01 Method, EN 15662 Method, and NY/T 1380-2007 Method procedures.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ceramic Homogenizers for 50 mL Tubes, 100/Pk.
50 mL Tubes
Ceramic Homogenizer