missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Ceramus™ Classic Fix

A universal bottle attachment dispenser for liquid handling.

Brand:  Hirschmann™ 9361100

 View more versions of this product

Product Code. 10518482

  • 636.00 € / Each
Estimated Shipment: 25-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Universal bottle attachment dispenser with fixed volume configuration
  • Certified conformity
  • With ceramic piston
  • Suitable for steam sterilization up to 121°C
  • Delivery complete with A 32 mm screw thread, 3 threaded adapters (A 28, S 40, A 45), 1 suction hose with union nut, 1 ejection unit and 1 fitting wrench
  • With individual serial number and calibration certificate
Specifications

Specifications

0.6%
0.2%
Screw thread, 3 threaded adapters, 1 suction hose, 1 ejection unit, 1 fitting wrench
Bottle Attachment Dispenser
50 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.