missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ Ceramus™ Fix Volume Bottle Top Dispensers
Guarantees consistent precision and volume dispensing with reproducible results independent of the user. The proven unit for manual dispensing, featuring superior-quality materials, simple handling and well-engineered user comfort to make laboratory routines easier.
502.00 € - 667.00 €
Specifications
Product Type | Bottletop Dispenser |
---|---|
Accuracy | 0.6% |
Autoclavable | Autoclavable |
Certifications/Compliance | ISO 8655 |
Description | ceramus™ FIX, fixed volume bottle top dispenser with reagent recirculation system |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10186050
|
Hirschmann™
9322100 |
2 fix |
502.00 €
Each |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||||
10702951
|
Hirschmann™
9332100 |
5 fix |
502.00 €
Each |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
10277910
|
Hirschmann™
9352100 |
25 fix |
667.00 €
Each |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
Description
- For fixed and variable volumes, from 0.2 mL to 60 mL
- Complete unit can be steam sterilized at 121°C without dismantling
- Free rotation
- Media recirculating system for simple and rapid venting without reagent loss
- Digital volume configuration engagement, calibrated for a lifetime
- Use within a temperature range of 4°C to 50°C
- Individual quality certificate with conformity certificate
Specifications
Bottletop Dispenser | |
Autoclavable | |
ceramus™ FIX, fixed volume bottle top dispenser with reagent recirculation system | |
Bottle | |
0.05 mL | |
1 Ea. |
0.6% | |
ISO 8655 | |
Aluminum Oxide Ceramic Piston, Duran Glass Cylinder | |
Thread adapter, dispensing nozzle, suction tube, valve wrench, calibration report | |
0.6% | |
0.2% |